Learn More
Research Products International Corp Mini-Blotting Box - Variety Pak, Black, 5 per Case
SDP
Catalog No. 50550306
Encompass
Includes one each of following sizes:
  • 5.4 x 4.1 x 2.0 cm
  • 7.2 x 5.0 x 2.5 cm
  • 9.0 x 6.4 x 2.1 cm
  • 11.5 x 8.9 x 2.2 cm
  • 15.4 x 10.4 x 2.5 cm
Catalog No. 50-550-306 Supplier Research Products International Corp Supplier No. 248767
Includes one each of following sizes:
  • 5.4 x 4.1 x 2.0 cm
  • 7.2 x 5.0 x 2.5 cm
  • 9.0 x 6.4 x 2.1 cm
  • 11.5 x 8.9 x 2.2 cm
  • 15.4 x 10.4 x 2.5 cm