Learn More
Research Products International Corp Mini-Blotting Containers, 7.2 x 5.0 x 2.5cm, Clear, 6 per Case
SDP
Catalog No. 50212597
Encompass
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50-212-597 Supplier Research Products International Corp Supplier No. 248715
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.