Learn More
Research Products International Corp Mini-Blotting Containers, 15.4 x 10.4 x 2.5cm, Black, 4 per Case
SDP
Catalog No. 50550382
Encompass
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50-550-382 Supplier Research Products International Corp Supplier No. 248718B
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.