Learn More
Research Products International Corp Mini-Blotting Containers, 18.75 x 12.75 x 2.0cm, Clear, 4 per Case
SDP
Catalog No. 50476690
Encompass
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50-476-690 Supplier Research Products International Corp Supplier No. 249601
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.