Learn More
Research Products International Corp Mini-Blotting Containers, 11.5 x 11.5 x 3.5cm, Clear, 6 per Case
SDP
Catalog No. 50476688
Encompass
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.
Catalog No. 50-476-688 Supplier Research Products International Corp Supplier No. 248714
Plastic containers for many laboratory applications. Containers have a smooth, flat surface that is ideal for blotting membranes or staining gels. Hinged lid with snap lock closure prevents evaporation of reagents. Constructed of ultra-clear polystyrene.