Learn More
Med Vet International Minibore Extension Set 6", J1317C, Each
Catalog No. 5000581304
Encompass_Preferred
Straight line - Female luerlock, proximal end - Male spin luerslip, distal end - Slide clamp Minibore Extension Set. 6" Priming volume 1.8ml
Catalog No. 50-005-81304 Supplier Med Vet International Supplier No. JOR1317C
Straight line - Female luerlock, proximal end - Male spin luerslip, distal end - Slide clamp Minibore Extension Set. 6" Priming volume 1.8ml