Learn More
Huber USA Minichiller 600 OLE
Catalog No. NC1827304
Encompass_Preferred
The compact Minichillers are an economic and environmentally friendly cooling solution for many laboratory applications. A calculation of water and waste water costs shows that a Minichiller can save up to 48,000 litres of water in a working week and due to its low procurement price, return on investment is achieved within a very short time.
Catalog No. NC1827304 Supplier Huber USA Supplier No. 3066.0001.98
Add to Cart
Add to Cart
Request a Quote
The compact Minichillers are an economic and environmentally friendly cooling solution for many laboratory applications. A calculation of water and waste water costs shows that a Minichiller can save up to 48,000 litres of water in a working week and due to its low procurement price, return on investment is achieved within a very short time.