Learn More
Huber USA Minichiller 300, Chiller
Catalog No. 501108696
Encompass_Preferred
‐ Temperature range: -20°C ... +40 (80)°C - Cooling capacity (15°C): 0,3 kW‐ Cooling capacity (0°C): 0,2 kW‐ Pressure pump max.: 14 l/min / 0,25 bar‐ Suction pump max. : 10,5 l/min / 0,17 bar‐ Power supply: 115V 1~ 60Hz
Catalog No. 50-110-8696 Supplier Huber USA Supplier No. 3006.0069.99
Add to Cart
Add to Cart
Request a Quote
‐ Temperature range: -20°C ... +40 (80)°C - Cooling capacity (15°C): 0,3 kW‐ Cooling capacity (0°C): 0,2 kW‐ Pressure pump max.: 14 l/min / 0,25 bar‐ Suction pump max. : 10,5 l/min / 0,17 bar‐ Power supply: 115V 1~ 60Hz