Learn More
Thomas Scientific MyFuge Mini Centrifuge, green lid 100-240V US Plug
SDP
Catalog No. 501458804
Encompass
The MyFuge™ Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6,000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. An increased capacity, eight-position microtube rotor is included and comes installed. Also included is a second rotor for 0.2ml PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use, storage in this compartment prevents loss or misplacement of the rotor. Clik-N-Lok™ INSTANT rotor attachment Eight position microtube rotor Storage compartment for PCR rotor & 0.5ml adapters Near silent operation Decelerates in just one second Four position PCR strip rotor
Catalog No. 50-145-8804 Supplier Thomas Scientific Supplier No. 1222U56
Add to Cart
Add to Cart
The MyFuge™ Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6,000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. An increased capacity, eight-position microtube rotor is included and comes installed. Also included is a second rotor for 0.2ml PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use, storage in this compartment prevents loss or misplacement of the rotor. Clik-N-Lok™ INSTANT rotor attachment Eight position microtube rotor Storage compartment for PCR rotor & 0.5ml adapters Near silent operation Decelerates in just one second Four position PCR strip rotor