Learn More
Stellar Scientific MiniGel Western Blot Box for Invitrogen/Novex mini-gels, Black, 10/CS
SDP
Catalog No. NC1171196
Encompass_Preferred
The latest addition to our line of Western Blot boxes, this light-safe, black-opaque box is perfect for fluorescent stains and all your mini-gel work. Designed for the Novex* system, but also compatible with other leading mini-gels. Each box features a smooth, flat surface to allow for complete gel coverage while sparing precious reagents.Dimensions are: 85 x 85 x 30mm Tight-fitting, easy to pull off lid Packaged in cases of 10 boxes*Novex is a registered trademark of ThermoFisher
Catalog No. NC1171196 Supplier Stellar Scientific Supplier No. MTC1300BK
The latest addition to our line of Western Blot boxes, this light-safe, black-opaque box is perfect for fluorescent stains and all your mini-gel work. Designed for the Novex* system, but also compatible with other leading mini-gels. Each box features a smooth, flat surface to allow for complete gel coverage while sparing precious reagents.Dimensions are: 85 x 85 x 30mm Tight-fitting, easy to pull off lid Packaged in cases of 10 boxes*Novex is a registered trademark of ThermoFisher