Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Biohazard Bag

Catalog No. 01824
Change view
Click to view available options
Dimensions (L x W):
6 x 9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
01-824 6 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-824 Supplier Minigrip™ Supplier No. SBL2R69B
Add to Cart
Add to Cart

Specifications

Thickness (English) 1.75 mil
Dimensions (L x W) 6 x 9 in.
Width (English) 6 in.
Height (English) 9 in.
Product Type 4-wall Biohazard Specimen Bag
Width (Metric) 15.2 cm
Thickness (Metric) 0.044 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.