Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Dressing Disposal Bags

Catalog No. 22310040
Change view
Click to view available options
Dimensions (L x W):
15 x 11 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-310-040 15 x 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-310-040 Supplier Minigrip™ Supplier No. UFDD1114
Add to Cart
Add to Cart

Ideal for non-sharps medical waste

  • Red bag with biohazard symbol
  • All bags are perforated for easy removal from the clip
  • Clip packs can be hung in the room or kept neatly in a drawer
  • Clip hook to hang bags on cart, wall, etc.

Order Info

WxH: 11 x 15 in.; 50 Bags/Clip, 10 Clips/Cs.

Specifications

For Use With (Application) Ideal for non sharps medical waste
Includes With biohazard symbol, hook (clip) to hang bags on cart, wall
Dimensions (L x W) 15 x 11 in.
Width (English) 11 in.
Height (English) 15 in.
Height (Metric) 38.1 cm
Product Type Dressing Disposal Bag
Width (Metric) 27.94 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.