Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Emesis Containment Bag

Catalog No. 22310011
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-310-011 10 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-310-011 Supplier Minigrip™ Supplier No. UFEMB1010
Add to Cart
Add to Cart

Special Emesis bag featuring two outside hand flaps and a liquid-tight safety seal

Perfect for bedside applications at surgery and oncology centers or emergency rooms and for odd-sized or specialty transport needs.

  • Extra-wide opening is designed for ease-of-use
  • Hand flaps on each side of the bag help to protect against splatter
  • Adhesive strip seals liquid tight, ready for safe disposal
  • Biohazard logo and illustrated instructions printed on the bag
  • A special score provides easy access for later analysis of content
  • Bag may be written on to identify contents
  • 100% disposable
  • Used by hospital personnel, patients, EMTs, paramedics, ambulance personnel, and nursing home staff

Specifications

Disposable Single-use
For Use With (Application) Hospital personnel, patients, EMT's paramedics, ambulance personnel and nursing home staff
Includes Liquid tight adhesive closure, score opening (for lab testing), biohazard symbol
Dimensions (L x W) 10 x 10 in.
Width (English) 10 in.
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Type Emesis Containment Bag
Width (Metric) 25.4 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.