Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Lab Guard™ Specimen Bags

Catalog No. 22306295
Change view
Click to view available options
Dimensions (L x W):
15 x 12 in. (38.1 x 30.5cm)
6 x 4 in. (15.2 x 10.1cm)
9 x 6 in. (23 x 15cm)
Thickness (English):
1.75 mil
2 mil
Product Type:
Biohazard Specimen Bag
Specimen Transport Bag
Quantity:
1000/Cs.
1000/Dispenser Bag
500/Dispenser Box
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Thickness (English) Product Type Quantity
22-306295 9 x 6 in. (23 x 15cm) 1.75 mil Biohazard Specimen Bag 500/Dispenser Box
22-310302 15 x 12 in. (38.1 x 30.5cm) 2 mil Biohazard Specimen Bag 1000/Cs.
01-823 9 x 6 in. (23 x 15cm) 1.75 mil Specimen Transport Bag 1000/Cs.
22-306-326 9 x 6 in. (23 x 15cm) 1.75 mil Biohazard Specimen Bag 1000/Dispenser Bag
22-306-325 6 x 4 in. (15.2 x 10.1cm) 1.75 mil Biohazard Specimen Bag 1000/Cs.
22-306297 9 x 6 in. (23 x 15cm) 1.75 mil Specimen Transport Bag 1000/Dispenser Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-306295 Supplier Minigrip™ Supplier No. SBL2X69B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect from inadvertent exposure to leakage and contamination

Bags with the destroyable biohazard symbol feature allow uncontaminated bag remains to be disposed in regular waste; if bag remains are contaminated, dispose in RED waste.

  • Zippered polyethylene three-wall bags with attached document compartment
  • Sample and paperwork stay together to reduce clerical errors and misidentification
  • TearZone™ EZ open feature means no leaks
  • Available with convenient DispenserBag™ and DispenserBox packaging

Order Info

Available plain or with orange and black biohazard labeling.†

Compliance

Comply with OSHA guidelines. STAT Specimen Transport Bags are FDA registered.

†Features a bright red requisition pocket and STAT labeling to easily identify urgent tests.

Specifications

Description 3-Walled Specimen Bag with Destroyable Biohazard Symbol, TearZone™
Closure Type Zip Seal
For Use With (Application) For transportation of specimens
Material Polyethylene
Quantity 500/Dispenser Box
Thickness (English) 1.75 mil
Dimensions (L x W) 9 x 6 in. (23 x 15cm)
Length (English) 9 in.
Width (English) 6 in.
Product Type Biohazard Specimen Bag
Length (Metric) 23 cm
Width (Metric) 15 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.