Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Lab Guard™ Specimen Bags with TearZone™ and Destroyable Biohazard Symbol

Catalog No. 22311106
Change view
Click to view available options
Color:
Green
Red
Red Tint
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-311-106 Green
22-311-105 -
22-311-107 Red
22-311-108 Red Tint
22-311-109 Yellow
22-311-111 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-311-106 Supplier Minigrip™ Supplier No. MGTZ69GRN
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Safe and durable specimen-handling reclosable bags feature open and close zipper technology and grip edges

With TearZone™ destroyable biohazard symbol feature.

  • Convenient 6 by 9 in. size
  • Color codes can be used to identify a specific urgency or department
  • TearZone™ feature will not leak — contains leak to one bag for overall cleanup and savings
  • Tears open easily at processing — no need for sharps to open the bag quickly
  • Grip edges make opening easier with gloved hands
  • Vivid biohazard risk warning graphic
  • Separate pouch for paperwork storage
  • Option to dispose unsoiled bags in regular waste means additional cost saving

Specifications

Color Green
For Use With (Application) Biohazard specimen transport
Includes TearZone™ and destroyable biohazard symbol
Material Virgin Polyethylene
Thickness (English) 2 mil
Dimensions (L x W) 9 x 6 in. (22.8 x 15.2 cm)
Length (English) 9 in.
Width (English) 6 in.
Product Type Biohazard Specimen Transport Bag
Length (Metric) 22.8 cm
Width (Metric) 15.2 cm
Thickness (Metric) 0.279 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.