Learn More
Minigrip™ Lab Guard™ Three Wall Biohazard Specimen Bags
Catalog No. 22311101
Change view
Click to view available options
Dimensions (L x W):
8 x 10 in.
Catalog No. Dimensions (L x W)
22-311-101 8 x 10 in.
1 options
Catalog No. 22-311-101 Supplier Minigrip™ Supplier No. SBL2X810B
Add to Cart
Add to Cart
Grip ridges make it easy to open and close even when wearing gloves
  • Vivid biohazard risk warning graphic
  • Satisfies OSHA Federal regulation 29CFR 1910.1030 Bloodborne Pathogens for containment and biohazard graphics
  • Meet NCCLS M29-T2 specimen handling guidelines
  • DispenserBox (500 count) bulk convenience packaging (for 8 x 10 in. bag)
Specifications
Thickness (English) 1.75 mil
Dimensions (L x W) 8 x 10 in.