Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Non-Biohazard Adhesive Closure Bags

Catalog No. 22310015
Change view
Click to view available options
Length (English):
10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-310-015 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-310-015 Supplier Minigrip™ Supplier No. UF95600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Feature the liquid-tight Press and Close™ adhesive closure system

  • Clear with black printing
  • Printed with instructions for use
  • No biohazard logo
  • Three-wall bags have separate document pocket

Specifications

Closure Type Self-adhesive
Color Clear
Length (English) 10 in.
Width (English) 6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.