Learn More
MSC Minigrip Reclosable Poly Bag, 13x18in, 6 Mil, 250/Case
Catalog No. 501917061
Encompass
Minigrip Reclosable Poly Bags are designed for secure storage and easy access, featuring a single-track, heavy-duty zipper that seals contents against moisture and contaminants. Constructed from premium, clear 6 mil polyethylene, these bags offer excellent puncture resistance and are suitable for repeated use.
  • Single track, heavy-duty zipper extends bag width
  • Keeps products clean and free from moisture
  • Premium poly offers superior clarity
  • Strong 6 mil construction provides puncture resistance
  • Reclosable design allows for multiple uses
  • Suitable for food contact, meeting FDA and USDA specifications
  • Inside usable dimensions: 13 x 18 in
Catalog No. 50-191-7061 Supplier MSC Supplier No. 39566096
Minigrip Reclosable Poly Bags are designed for secure storage and easy access, featuring a single-track, heavy-duty zipper that seals contents against moisture and contaminants. Constructed from premium, clear 6 mil polyethylene, these bags offer excellent puncture resistance and are suitable for repeated use.
  • Single track, heavy-duty zipper extends bag width
  • Keeps products clean and free from moisture
  • Premium poly offers superior clarity
  • Strong 6 mil construction provides puncture resistance
  • Reclosable design allows for multiple uses
  • Suitable for food contact, meeting FDA and USDA specifications
  • Inside usable dimensions: 13 x 18 in