Learn More
Minigrip™ Red Line™ Plain Reclosable Zipper Bags with White Block
Catalog No. 22306379
Change view
Click to view available options
Dimensions (L x W):
2 x 3 in.
9 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-306-379 9 x 12 in.
22-306-372 2 x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-306-379 Supplier Minigrip™ Supplier No. MGRL2W0912
Add to Cart
Add to Cart
Reinforced zipper and side seams to keep bags from splitting when opened or filled
  • Premium zipper easily snaps open and zips securely shut
  • Superior clarity
  • Packaged in convenient DispenserBag™ packaging
  • Remove a bag at a time through the mid package opening or through the zipper at the top
  • Packaged in high quality corrugated boxes for reliable storage and easy handling
  • Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
Specifications
Closure Type Reinforced Zipper and Side Seams
Material LDPE, LLDPE
Thickness (English) 2 mil
Dimensions (L x W) 9 x 12 in.