Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ Reloc ZIPPIT™ Clear Non-Biohazard Reclosable Zipper Bags

Catalog No. 22310025
Change view
Click to view available options
Dimensions (L x W):
10 x 8 in. (25.4 x 20.3cm)
12 x 9 in. (30.5 x 22.9cm)
15 x 12 in. (38.1 x 30.5cm)
18 x 13 in. (45.7 x 33cm)
3 x 2 in. (7.6 x 5cm)
3 x 3 in. (7.6 x 7.6cm)
5 x 3 in. (12.7 x 7.6cm)
6 x 4 in. (15.2 x 10.2cm)
7 x 5 in. (17.8 x 12.7cm)
8 x 2 in. (20.3 x 5cm)
8 x 6 in. (20.3 x 15.2cm)
Thickness (English):
2 mil
4 mil
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Thickness (English)
22-310-025 8 x 6 in. (20.3 x 15.2cm) 2 mil
22-310-029 15 x 12 in. (38.1 x 30.5cm) 4 mil
22-310-023 6 x 4 in. (15.2 x 10.2cm) 4 mil
22-310-028 15 x 12 in. (38.1 x 30.5cm) 2 mil
22-310-030 18 x 13 in. (45.7 x 33cm) 2 mil
22-310-022 6 x 4 in. (15.2 x 10.2cm) 2 mil
22-310-019 3 x 3 in. (7.6 x 7.6cm) 2 mil
22-310-020 5 x 3 in. (12.7 x 7.6cm) 2 mil
22-310-024 7 x 5 in. (17.8 x 12.7cm) 2 mil
22-310-026 10 x 8 in. (25.4 x 20.3cm) 2 mil
22-310-027 12 x 9 in. (30.5 x 22.9cm) 2 mil
22-306-310 3 x 2 in. (7.6 x 5cm) 2 mil
22-306-330 8 x 2 in. (20.3 x 5cm) 2 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 22-310-025 Supplier Minigrip™ Supplier No. MGZ2P0608
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Economical bags with zipper closure

  • Clear (plain) bags with no biohazard logo
  • Wide range of sizes and gauges
  • With or without white block and hangholes
  • DispenserBag™ convenience packaging

To avoid the risk of cross-contamination, specimen bags should not be reused.

Specifications

Closure Type Zip Seal
Color Clear
Thickness (English) 2 mil
Dimensions (L x W) 8 x 6 in. (20.3 x 15.2cm)
Length (English) 8 in.
Width (English) 6 in.
Product Type Specimen Transport Bag
Length (Metric) 20.3 cm
Width (Metric) 15.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.