Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-GARD™ STAT Specimen Transport Bag

Catalog No. 018194
Change view
Click to view available options
Color:
Red
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-819-4 Yellow
22-307-949 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-819-4 Supplier Minigrip™ Supplier No. UF95600STB
Add to Cart
Add to Cart

Easily identifies specimens for priority handling; ideal for pneumatic tube systems

  • Double pocket, three-wall bag features Press-and Close™ one-touch safety seal for specimens and integral outside requisition pocket
  • Prominent “STAT” copy displayed on bag with biohazard symbol
  • Seals in specimens to reduce the risk of contact with dangerous body fluids
  • Not reusable; once sealed, cannot be reopened without compromising the integrity of the bag

Order Info

Dimensions: 6W x 10 in. H. Case of 1000 in packs of 100.

Compliance

Meets OSHA and NCCLS requirements for liquid-tight transport

Specifications

Color Yellow
Disposable Single-use
For Use With (Application) For specimen transport
Includes Press and Close™ liquid tight closure printed with biohazard logo and instructions for use
Width (English) 6 in.
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Type Specimen Transport Bag
Width (Metric) 15.24 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.