Learn More
Minigrip™ SPECI-GUARD™ Nonbiohazard Specimen Bags
Catalog No. 22311206
Change view
Click to view available options
Dimensions (L x W):
6 x 6 in. (15.2 x 15.5 cm)
Catalog No. Dimensions (L x W)
22-311-206 6 x 6 in. (15.2 x 15.5 cm)
1 options
Catalog No. 22-311-206 Supplier Minigrip™ Supplier No. UF9566SP
Add to Cart
Add to Cart
Ideal when requisitions are not used, in diagnostic kits, for single urine cups or for single/multiple tubes
  • Disposable
  • Double pocket bag
  • Separate outside pocket for requisition slip
  • With Press and Close™ liquid-tight closure
  • Printed with instructions for use
  • Remains closed if air is trapped inside
Specifications
Dimensions (L x W) 6 x 6 in. (15.2 x 15.5 cm)
Product Type Specimen Bag