Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-ZIP™ Non-Biohazard Reclosable Clear Bags

Catalog No. 22310414
Change view
Click to view available options
Length (English):
9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-310-414 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-310-414 Supplier Minigrip™ Supplier No. IP69CP
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Non-printed, clear reclosable bags

  • Three walls, including information pocket
  • Tear line
  • Bilingual in English and Spanish
  • W x H: 6 in. x 9 in.

Order Info

Case of 1000 supplied in packs of 100.

Specifications

Color Clear
Length (English) 9 in.
Width (English) 6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.