Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-ZIP™ Reclosable Clear Biohazard Bags with Black and Orange Biohazard Warning

Catalog No. 22310404
Change view
Click to view available options
Width (English):
4 in.
6 in.
8 in.
12 in.
20 in.
Height (English):
6 in.
9 in.
10 in.
15 in.
24 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English)
22-310-404 8 in. 10 in.
22-310-402 4 in. 6 in.
22-310-403 6 in. 9 in.
22-310-405 12 in. 15 in.
22-310-406 20 in. 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-310-404 Supplier Minigrip™ Supplier No. IP810B3T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed with temperature indicators and tear line

  • Double pocket, three-wall bags
  • Bilingual in English and Spanish

Order Info

Case of 1000 supplied in packs of 100.

Specifications

Color Clear
For Use With (Application) Easy-to-open tear line
Includes Black and orange biohazard warning, double pocket, temperature indicators and tear line
Width (English) 8 in.
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Type Reclosable Biohazard Bag
Width (Metric) 20.32 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.