Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ SPECI-ZIP™ Reclosable Red Biohazard Bags

Catalog No. 22310410
Change view
Click to view available options
Width (English):
6 in.
8 in.
Height (English):
9 in.
10 in.
Color:
Red
Transparent Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Height (English) Color
22-310-410 8 in. 10 in. Red
22-310-409 6 in. 9 in. -
22-310-096 6 in. 9 in. Transparent Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-310-410 Supplier Minigrip™ Supplier No. IP810RSTAT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With black STAT and biohazard warning

  • Designed with info pocket (three wall) and tear line
  • Bilingual in English and Spanish

Order Info

Case of 1000 in packs of 100.

Specifications

Color Red
For Use With (Application) Designed with info pocket, temperature indicators and tear line
Includes Black STAT/ biohazard warning
Width (English) 8 in.
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Type Reclosable Biohazard Bag
Width (Metric) 20.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.