Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ StaticGuard™ Anti-Static Bags

Catalog No. 22311221
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in. (30.48 x 30.48cm)
3 x 5 in. (7.62 x 12.70cm)
4 x 6 in. (10.16 x 15.24cm)
6 x 8 in. (15.24 x 20.32cm)
6 x 9 in. (15.24 x 22.86cm)
8 x 10 in. (20.32 x 25.40cm)
Quantity:
1000/Pk.
500/Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Quantity
22-311-221 3 x 5 in. (7.62 x 12.70cm) 1000/Pk.
22-311-224 6 x 9 in. (15.24 x 22.86cm) 1000/Pk.
22-311-222 4 x 6 in. (10.16 x 15.24cm) 1000/Pk.
22-311-223 6 x 8 in. (15.24 x 20.32cm) 1000/Pk.
22-311-225 8 x 10 in. (20.32 x 25.40cm) 1000/Pk.
22-311-227 12 x 12 in. (30.48 x 30.48cm) 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-311-221 Supplier Minigrip™ Supplier No. MGST4P0305
Add to Cart
Add to Cart

For transportation of electronic components that are prone to damage caused by electrostatic discharge

  • Nonbiohazard
  • With zipper
  • Thickness of 4 mil
  • Pink
  • Available in different sizes

Specifications

Description Anti-Static Bag
Closure Type Zip Seal
Color Pink
Quantity 1000/Pk.
Thickness (English) 4 mil
Dimensions (L x W) 3 x 5 in. (7.62 x 12.70cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.