Learn More
REPLIGEN CORPORATION MINIKROS SAMPLER 100 KDA 0.5 M
SDP
Supplier:  REPLIGEN CORPORATION S04E10005S
Encompass
Minikros Sampler 100 kDa 0.5 mm fiber, mPES 1600 cm2 surface area hollow fiber
Catalog No. NC2108454