Learn More
Electron Microscopy Sciences miniLIGHT Set 4 2W
SDP
Catalog No. 5034628
Encompass_Preferred
Constructed from the highest quality material and small as its name, this miniLight offers the most universal light source, handling many tasks that require a bright light but with a small foot print that fits into most locations.
The miniLight's main component is the miniLamp. This is a small but powerful light source. It shines very bright on a small area. It has a low dissipation loss with no hot part. Long life because the lifetime of a LED is 50,000 hours. Little power needed (1watt). Adjustable intensity and CE compatible.
Catalog No. 50-346-28 Supplier Electron Microscopy Sciences Supplier No. 7166604
Constructed from the highest quality material and small as its name, this miniLight offers the most universal light source, handling many tasks that require a bright light but with a small foot print that fits into most locations.
The miniLight's main component is the miniLamp. This is a small but powerful light source. It shines very bright on a small area. It has a low dissipation loss with no hot part. Long life because the lifetime of a LED is 50,000 hours. Little power needed (1watt). Adjustable intensity and CE compatible.