Learn More
Bertin Corp MINILYS HOMOGENIZER
SDP
Catalog No. 501542929
Encompass_Preferred
Minilys homogenizer is a compact and flexible instrument for sample preparation. Specially designed to offer an optimal efficiency on every kind of samples. Minilys is able to handle 3 tubes of 2mL/0.5mL or 1 tube of 7mL. Its manual locking system makes Minilys an easy pick up laboratory instrument.
Catalog No. 50-154-2929 Supplier Bertin Corp Supplier No. P000673MLYS0A.0
Minilys homogenizer is a compact and flexible instrument for sample preparation. Specially designed to offer an optimal efficiency on every kind of samples. Minilys is able to handle 3 tubes of 2mL/0.5mL or 1 tube of 7mL. Its manual locking system makes Minilys an easy pick up laboratory instrument.