Learn More
Alkali Scientific Alkali Scientific™ MiniMag S1005 Mini Magnetic Stirrer, 115V
SDP
Catalog No. 503029588
Encompass_Preferred
“Mini” sized, less than 3.5″ wide Adjustable speed control, 100 to 1500rpm Effectively mixes volumes up to 1.5L With a square footprint of under 5 inches, the Alkali Scientific™ S1005  MiniMag™ is the most compact stirrer in the Alkali line. Despite its modest dimensions, the 3.4 inch square work surface accepts a wide variety of beakers, flasks, bottles and other laboratory vessels for stirring volumes as large as 1.5 liters. The Magnetic Stirrer speed is continuously adjustable from 100 to 1500rpm. For temperature sensitive samples, the MiniMag is incubator and cold-room safe for use from +4°C to +45°C.
Catalog No. 50-302-9588 Supplier Alkali Scientific Supplier No. R3760MM
“Mini” sized, less than 3.5″ wide Adjustable speed control, 100 to 1500rpm Effectively mixes volumes up to 1.5L With a square footprint of under 5 inches, the Alkali Scientific™ S1005  MiniMag™ is the most compact stirrer in the Alkali line. Despite its modest dimensions, the 3.4 inch square work surface accepts a wide variety of beakers, flasks, bottles and other laboratory vessels for stirring volumes as large as 1.5 liters. The Magnetic Stirrer speed is continuously adjustable from 100 to 1500rpm. For temperature sensitive samples, the MiniMag is incubator and cold-room safe for use from +4°C to +45°C.