Learn More
MOLTOX Molecular Toxicology Inc Minimal Glucose Agar 0.4% Glucose w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.
SDP
Catalog No. NC3432876
Encompass
Minimal Glucose Agar 0.4% Glucose w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.
Catalog No. NC3432876 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S314.10
Minimal Glucose Agar 0.4% Glucose w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.