Learn More
MOLTOX Molecular Toxicology Inc Minimal Glucose Agar w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.
SDP
Catalog No. NC3432875
Encompass
Minimal Glucose Agar w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.
Catalog No. NC3432875 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S313.10
Minimal Glucose Agar w/ L-Histidine & D-Biotin 6 well plate. 5 mL/well. 10 plates/sleeve. Store at room temperature.