Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium Eagle (MEM) Cell Culture Medium | 10 L
Catalog No. NC2283616
Encompass
Minimum Essential Medium Eagle (MEM), cell culture medium | Mol Wt: N/A | CAS No: N/A | N/A | 10LThis dry powder basal medium must be reconstituted with sodium bicarbonate and sterile water prior to use. It contains essential amino acids, vitamins, inorganic salts, glucose and other nutrients to support mammalian cell growth when supplemented with serum. Suitable for culturing various adherent and suspension cell lines. Provided as a 10 liter format for large scale mammalian cell culture applications.
This item is not returnable. View return policy
Catalog No. NC2283616 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 56416C10L
Minimum Essential Medium Eagle (MEM), cell culture medium | Mol Wt: N/A | CAS No: N/A | N/A | 10LThis dry powder basal medium must be reconstituted with sodium bicarbonate and sterile water prior to use. It contains essential amino acids, vitamins, inorganic salts, glucose and other nutrients to support mammalian cell growth when supplemented with serum. Suitable for culturing various adherent and suspension cell lines. Provided as a 10 liter format for large scale mammalian cell culture applications.