Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium E
Catalog No. 502900056
Encompass
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids some at increased concentrations. In addition it contains 5 additional vitamins.
Catalog No. 50-290-0056 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M089410L
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids some at increased concentrations. In addition it contains 5 additional vitamins.