Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium E
Catalog No. 502901893
Encompass
It contains 19 amino acids. The essential amino acids and the non-essential amino acids L-ala L-asn L-asp L-glu L-gly L-pro and L-ser. May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
Catalog No. 50-290-1893 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M064350L
It contains 19 amino acids. The essential amino acids and the non-essential amino acids L-ala L-asn L-asp L-glu L-gly L-pro and L-ser. May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.