Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium Eagle, 500 mL
Catalog No. NC1800349
Encompass
Contains 19 amino acids. The essential amino acids and the non-essential amino acids; L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser. May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
Catalog No. NC1800349 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M5650500ML
Contains 19 amino acids. The essential amino acids and the non-essential amino acids; L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser. May be preferred for growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.