Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium Eagle With Earle's salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC2042664
Encompass
Minimum Essential Medium Eagle With Earle's salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC2042664 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M2279500ML
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Minimum Essential Medium Eagle With Earle's salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | 500ML