Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential Medium Eagle With Hanks' salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L
Catalog No. NC1526905
Encompass
Minimum Essential Medium Eagle With Hanks' salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L
Catalog No. NC1526905 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M101810L
Minimum Essential Medium Eagle With Hanks' salts, L-glutamine and non-essential amino acids, without sodium bicarbonate, powder, suitable for cell culture | 10L