Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential medum E500ML
Catalog No. 502914204
Encompass
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids some at increased concentrations. In addition it contains 5 additional vitamins.
Catalog No. 50-291-4204 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M452624X500ML
This is the most enriched variation of the MEM formulation offered. It contains all 21 normal amino acids some at increased concentrations. In addition it contains 5 additional vitamins.