Learn More
Sigma Aldrich Fine Chemicals Biosciences Minimum Essential medum E500ML
Catalog No. 502901674
Encompass
Modification of the original MEM formulation. Earles salts have been replaced with Hanks salts. Since the bicarbonate concentration is substantially lower this medium has a substantially reduced buffering capacity.
Catalog No. 50-290-1674 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M4780500ML
Modification of the original MEM formulation. Earles salts have been replaced with Hanks salts. Since the bicarbonate concentration is substantially lower this medium has a substantially reduced buffering capacity.