Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minioxidil, 98.2%, MP Biomedicals™

Catalog No. ICN15571980 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15571980 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15571980 Supplier MP Biomedicals, Inc Supplier No. 0215571980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.253
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
Synonym minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
PubChem CID 4201
IUPAC Name 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O

Specifications

Color White
Assay Percent Range 0.982
Quantity 100 mg
Formula Weight 209.3
Physical Form Crystalline Powder
Chemical Name or Material Minioxidil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.