Learn More
AMPLYUS LLC miniPCR bio miniPCR Lab in a Box™ Kit #4 (mini16X upgrade)
SDP
Catalog No. NC3597534
Encompass_Preferred
Lab in a Box #4: a biotech classroom on wheels. Includes 4 miniPCR mini16X thermal cyclers, 4 blueGel™ electrophoresis systems, a protective rigid carrying case (24 x 17 x 10 in), 8 micropipettes (2-20 µl), 1 micropipette (20-200 µl), and 1 micropipette (1-10 µl).
Catalog No. NC3597534 Supplier AMPLYUS LLC Supplier No. LB500016
Add to Cart
Add to Cart
Request a Quote
Lab in a Box #4: a biotech classroom on wheels. Includes 4 miniPCR mini16X thermal cyclers, 4 blueGel™ electrophoresis systems, a protective rigid carrying case (24 x 17 x 10 in), 8 micropipettes (2-20 µl), 1 micropipette (20-200 µl), and 1 micropipette (1-10 µl).