Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ MiniPet™ Pipette for Cholesterol Analyzers

Catalog No. 23763155 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Capacity (Metric):
40 μL
Sample Type:
Blood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sample Type
23-763-155 40 μL Blood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-763-155 Supplier Abbott™ Supplier No. 13014
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For use with the Cholestech LDX™ System.

For use with Cholestech LDX lipid monitoring system.

For use with Cholestech LDX lipid monitoring system.

Specifications

Type MiniPet™ Pipette
Sample Type Blood
For Use With (Equipment) Cholestech LDX™ lipid monitoring system
Capacity (Metric) 40 μL
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.