Learn More
AMPLYUS LLC miniPCR bio miniRack for 8 PCR tubes (0.2 ml), green
SDP
Catalog No. 502237969
Encompass_Preferred
Ideal tube rack to satisfy all pre-PCR setup or post-PCR sampling applications. Holds individual PCR tubes or 8-tube strips. Injection-molded durable design. Green.
Catalog No. 50-223-7969 Supplier AMPLYUS LLC Supplier No. CM100302
Ideal tube rack to satisfy all pre-PCR setup or post-PCR sampling applications. Holds individual PCR tubes or 8-tube strips. Injection-molded durable design. Green.