Learn More
BURKLE INC MiniSampler PTFE, complete
Catalog No. 5000025966
Encompass_Preferred
With the vacuum sampler MiniSampler, the sample liquid only comes into contact with the suction hose and the original sample bottle. The possibility of cross-contamination is greatly reduced. Immediately after taking the sample, the bottles are sealed. The disposable suction hose can quickly be replaced. This guarantees that the samples are never contaminated. If necessary, a new hose can be used for every single sample.
Due to its small diameter (8 mm (0.31 in.)), the hose can reach narrow, seemingly inaccessible sampling areas.
Catalog No. 50-000-25966 Supplier BURKLE INC Supplier No. 53050110
With the vacuum sampler MiniSampler, the sample liquid only comes into contact with the suction hose and the original sample bottle. The possibility of cross-contamination is greatly reduced. Immediately after taking the sample, the bottles are sealed. The disposable suction hose can quickly be replaced. This guarantees that the samples are never contaminated. If necessary, a new hose can be used for every single sample.
Due to its small diameter (8 mm (0.31 in.)), the hose can reach narrow, seemingly inaccessible sampling areas.