Learn More
Huber USA Ministat 125, Refrigerated Heating Circulator Bath
Catalog No. 501108704
Encompass_Preferred
‐ Temperature range: -25°C ... +150°C - Cooling capacity (15°C): 0,3 kW‐ Cooling capacity (0°C): 0,21 kW‐ Heating capacity: 1 kW‐ Pressure pump max.: 22 l/min / 0,7 bar‐ Suction pump max. : 16 l/min / 0,4 bar‐ Pump connection: M16x1 male ‐ Power supply: 115V 1~ 60Hz
Catalog No. 50-110-8704 Supplier Huber USA Supplier No. 2014.0012.01
Add to Cart
Add to Cart
Request a Quote
‐ Temperature range: -25°C ... +150°C - Cooling capacity (15°C): 0,3 kW‐ Cooling capacity (0°C): 0,21 kW‐ Heating capacity: 1 kW‐ Pressure pump max.: 22 l/min / 0,7 bar‐ Suction pump max. : 16 l/min / 0,4 bar‐ Pump connection: M16x1 male ‐ Power supply: 115V 1~ 60Hz