Learn More
Sirius Automation Inc MiniTasker Benchtop Lab Robot
SDP
Catalog No. 5000078211
Encompass
Process everyday tasks such as Analytical Weighing (4 or 5 place) Sample ID (1D or 2D barcodes) Sorting Dilutions Vortex Mixing Internal Standard Addition Standard Prep Un-Capping Re-Capping Transfers Aliquoting Large Sample Capacity (max 20 microplate racks or custom equivalent) choice of disposables and labware yet benchtop compact (3 x2 ) footprint and easy to use interface (21CFR11 compliance options) with LIMS compatible data management The robot hardware is exceptionally fast and incorporates a built-in enclosure and optional hardware interlock to ensure safety Each robotic step is recorded and logged such that there is a unique and forensic sample history to be readily maintained User LIMS interfaces are compatible with 21CFR11 GLP compliance The whole unit requires just over 3 feet of linear bench space and is sufficiently stable to be mounted on wheeled carts if lab-to-lab portability is required
Catalog No. 50-000-78211 Supplier Sirius Automation Inc Supplier No. MNT70101
Add to Cart
Add to Cart
Request a Quote
Process everyday tasks such as Analytical Weighing (4 or 5 place) Sample ID (1D or 2D barcodes) Sorting Dilutions Vortex Mixing Internal Standard Addition Standard Prep Un-Capping Re-Capping Transfers Aliquoting Large Sample Capacity (max 20 microplate racks or custom equivalent) choice of disposables and labware yet benchtop compact (3 x2 ) footprint and easy to use interface (21CFR11 compliance options) with LIMS compatible data management The robot hardware is exceptionally fast and incorporates a built-in enclosure and optional hardware interlock to ensure safety Each robotic step is recorded and logged such that there is a unique and forensic sample history to be readily maintained User LIMS interfaces are compatible with 21CFR11 GLP compliance The whole unit requires just over 3 feet of linear bench space and is sufficiently stable to be mounted on wheeled carts if lab-to-lab portability is required