Learn More
World Precision Instrument MiniTip - Fiber Optic Oxygen Sensor
SDP
Catalog No. 502521411
Encompass
This oxygen dipping probe has a tip diameter of 4 mm and consists of a polymer optical fiber, with an oxygen sensitive coating. The MiniTip's range is 0 to 100%. This robust sensor has a response time (t90) of approximately 40 s.
Catalog No. 50-252-1411 Supplier World Precision Instrument Supplier No. 501641
This oxygen dipping probe has a tip diameter of 4 mm and consists of a polymer optical fiber, with an oxygen sensitive coating. The MiniTip's range is 0 to 100%. This robust sensor has a response time (t90) of approximately 40 s.