Learn More
Hoefer, Inc Hoefer™ SE300 miniVE Blotting Unit
SDP
Catalog No. NC2740673
Encompass_Preferred
The SE300 minVE is a sturdy, compact vertical protein electrophoresis unit that also performs electro transfer (blotting).Blot Module Includes:Blot ModuleBlotter Paper, 25 sheetsDacron Sponges, 6 mm thick, 4 pcs.
Catalog No. NC2740673 Supplier Hoefer, Inc Supplier No. SE302
The SE300 minVE is a sturdy, compact vertical protein electrophoresis unit that also performs electro transfer (blotting).Blot Module Includes:Blot ModuleBlotter Paper, 25 sheetsDacron Sponges, 6 mm thick, 4 pcs.