Learn More
Braintree Scientific Minivette POCT, 20ul, Neutral, white, 200/pk
SDP
Catalog No. 501955132
Encompass
Minivette POCT - Easy sample recovery due to optimal capillary features. Precise dispensing of small whole blood volumes. Prevents spillage during transfer to test card. Available in 20ul and 50ul volumes. Preparations: Neutral, heparin and EDTA **Sold for research purposes only and not for human use**
Catalog No. 50-195-5132 Supplier Braintree Scientific Supplier No. MP202111PK
 Restricted Item
Add to Cart
Add to Cart
Minivette POCT - Easy sample recovery due to optimal capillary features. Precise dispensing of small whole blood volumes. Prevents spillage during transfer to test card. Available in 20ul and 50ul volumes. Preparations: Neutral, heparin and EDTA **Sold for research purposes only and not for human use**